Learn More
Methyllithium, 1.6M sol. in diethyl ether (± 5% w/v), AcroSeal™, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless to Yellow |
Linear Formula | CH3Li |
Fieser | 01,686; 02,274; 03,202; 05,448; 06,384; 07,242; 08,342; 09,311; 11,340; 12,324; 16,219 |
Density | 0.7000g/mL |
Solubility Information | Solubility in water: reacts. |
IUPAC Name | lithium;carbanide |
Formula Weight | 21.96 |
Percent Purity | 4.5 to 5.5% (as MeLi) |
Packaging | AcroSeal™ Glass bottle |
Specific Gravity | 0.7 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0461091 | Certificate of Analysis | 21/03/2024 | 188758000,188751000 |
A0417818 | Certificate of Analysis | 11/03/2024 | 188758000,188751000 |
A0451332 | Certificate of Analysis | 15/01/2024 | 188758000,188751000 |
A0439116 | Certificate of Analysis | 06/10/2023 | 188758000,188751000 |
A0389709 | Certificate of Analysis | 12/12/2022 | 188758000,188751000 |
Safety and Handling



- Methyllithium, 1.6 M solution in diethyl ether
- Danger
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
- Flammable liquid Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H224-Extremely flammable liquid and vapour.
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P402+P404-Store in a dry place. Store in a closed container.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- MIXTURE LIST-Contains : Lithium, methyl-, Ethyl ether
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.