missing translation for 'onlineSavingsMsg'
Learn More
Methyl (R)-(-)-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals Product Code.: 11379005

Methyl (R)-(-)-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals

Product Code. 11379005
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11379005

Brand: Thermo Scientific Alfa Aesar L14151.06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can undergo aldol-type reactions with aldehydes with either syn- or anti-selectivity according to the conditions, for use in the synthesis of chiral β-lactams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Can undergo aldol-type reactions with aldehydes with either syn- or anti-selectivity according to the conditions, for use in the synthesis of chiral ß-lactams.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3976-69-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00063289
InChI Key LDLDJEAVRNAEBW-SCSAIBSYSA-N
Synonym methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester
PubChem CID 2724279
IUPAC Name methyl (3R)-3-hydroxybutanoate
SMILES CC(CC(=O)OC)O

Specifications

Density 1.055
Boiling Point 56°C to 58°C (11 mmHg)
Flash Point 73°C (163°F)
Odor Pleasant
Refractive Index 1.422
Quantity 5 g
Beilstein 3648161
Solubility Information Slightly soluble in water.
Optical Rotation −48.5° (c=1.3 in Chloroform)
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material Methyl (R)-(-)-3-hydroxybutyrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl (R)-(-)-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.