missing translation for 'onlineSavingsMsg'
Learn More
Methanol-d{4}, 100%(Isotopic), Thermo Scientific Chemicals Product Code.: 11354378

Methanol-d{4}, 100%(Isotopic), Thermo Scientific Chemicals

Product Code. 11354378
10 Ea.
Click to view available options
Quantity:
10 Ea.
2 Ea.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11354378

Brand: Thermo Scientific Alfa Aesar 042320.KV

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Methanol-d_4 is the labeled analog of methyl alcohol, a common laboratory solvent used primarily in spectroscopic techniques (UV/VIS, NMR, IR).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811-98-3
Molecular Formula CH4O
Molecular Weight (g/mol) 36.066
MDL Number MFCD00044637
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % dShow MoreShow Less
PubChem CID 71568
IUPAC Name trideuterio(deuteriooxy)methane
SMILES CO

Specifications

Physical Form Liquid
Color Colorless
Density 0.89 g/mL
Flash Point 11°C (52°F)
Odor Mild
Packaging Prescored Ampule
Quantity 10 Ea.
Melting Point -98°C
Boiling Point 64.7°C
UN Number UN1230
Solubility Information Fully miscible in water.
Formula Weight 36.07
Percent Purity 100%
Chemical Name or Material Methanol-d4, Isotopic
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP03I037 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number042320.KV,042320.SA
Lot NumberN16I035 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number042320.KV,042320.SA
Lot NumberY01H070 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number042320.KV,042320.SA
Lot NumberS25E035 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042320.KV,042320.SA
Lot NumberL15P56 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042320.KV,042320.SA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol-d{4}, 100%(Isotopic), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.