Learn More
Eppendorf™ Metering, For Varitip S Dispensing Part
GREENER_CHOICE
Aspirate liquid from tall, narrow-necked vessels. Eppendorf™ Varitip™ S Forms a system with the Eppendorf Maxitip for aspirating 2.5 to 10mL with Eppendorf Varipette.
Brand:  Eppendorf™ 0030050533
92.70 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10603381
  • 93.80€ / Pack of 30
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of highest quality, virgin polypropylene (PP) free of biocides, plasticizers and latex
  • Available in packs of 30
  • Air displacement principle
  • No carryover
  • Ensures drip-free dispensing of liquids with a high vapor pressure
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Eppendorf Maxitip and Varipette
30 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ Metering, For Varitip S Dispensing Part > Metering, For Varitip S Dispensing Part