missing translation for 'onlineSavingsMsg'
Learn More
Metalaxyl in Methanol 100μg/mL, Fisher Chemical™ Product Code.: 12909903
compliance-icons compliance-icons compliance-icons

Metalaxyl in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12909903
1 mL
Click to view available options
Quantity:
1 mL
This item is not returnable. View return policy

Product Code. 12909903

Brand: Fisher Chemical P831230

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 100μg/mL
CAS 57837-19-1
Packaging Ampoule(s)
Quantity 1 mL
Molecular Formula C15H21NO4
Solvent or Matrix Methanol
UN Number 1230
InChI Key ZQEIXNIJLIKNTD-UHFFFAOYSA-N
SMILES CC1=C(C(=CC=C1)C)N(C(C)C(=O)OC)C(=O)COC
IUPAC Name methyl 2-(N-(2-methoxyacetyl)-2,6-dimethylanilino)propanoate
Molecular Weight (g/mol) 279.336
PubChem CID 42586
ChEBI CHEBI:82790
Chemical Name or Material Metalaxyl
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1086804 Certificate TypeCertificate of Analysis Date05/03/2025 Catalog NumberP831230
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution Metalaxyl 100ug/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metalaxyl in Methanol 100μg/mL, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.