missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Mercury Free COD Test Tubes
153.00€
Specifications
Description | Mercury Free COD Test Tube |
---|---|
For Use With (Application) | For COD Determination |
Quantity | 25 Pack |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11702435
|
Hanna Instruments™
HI93754D-25 |
0 to 150 mg/L |
153.00€
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11722435
|
Hanna Instruments™
HI93754E-25 |
0 to 15,000 mg/L |
153.00€
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Mercury Free COD Test Tube | |
For COD Determination | |
25 Pack |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- X25 TUBE TEST COD 0-150MG/L WH HG
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P391-Collect spillage.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, silver sulphate, potassium dichromate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hanna Instruments™ Mercury Free COD Test Tubes