Learn More
Melphalan, 94%, Thermo Scientific Chemicals
Melphalan, CAS # 148-82-3, belongs to the class of nitrogen mustards with anti-neoplastic activity.
176.00€ - 575.00€
Chemical Identifiers
CAS | 148-82-3 |
---|---|
Molecular Formula | C13H18Cl2N2O2 |
Molecular Weight (g/mol) | 305.2 |
InChI Key | SGDBTWWWUNNDEQ-LBPRGKRZSA-N |
Synonym | melphalan, alkeran, melfalan, phenylalanine mustard, l-sarcolysine, l-pam, l-sarcolysin, l-phenylalanine mustard, levofalan, p-l-sarcolysin |
PubChem CID | 460612 |
ChEBI | CHEBI:28876 |
IUPAC Name | (2S)-2-amino-3-[4-[bis(2-chloroethyl)amino]phenyl]propanoic acid |
SMILES | C1=CC(=CC=C1CC(C(=O)O)N)N(CCCl)CCCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15531033
|
Thermo Scientific Acros
458650010 |
1 g |
176.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15572343
|
Thermo Scientific Acros
458650050 |
5 g |
575.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
148-82-3 | |
305.2 | |
melphalan, alkeran, melfalan, phenylalanine mustard, l-sarcolysine, l-pam, l-sarcolysin, l-phenylalanine mustard, levofalan, p-l-sarcolysin | |
CHEBI:28876 | |
C1=CC(=CC=C1CC(C(=O)O)N)N(CCCl)CCCl |
C13H18Cl2N2O2 | |
SGDBTWWWUNNDEQ-LBPRGKRZSA-N | |
460612 | |
(2S)-2-amino-3-[4-[bis(2-chloroethyl)amino]phenyl]propanoic acid |
Specifications
148-82-3 | |
Authentic | |
Glass Bottle | |
(ClCH2CH2)2NC6H4CH2CH(NH2)COOH | |
SGDBTWWWUNNDEQ-LBPRGKRZSA-N | |
(2S)-2-amino-3-[4-[bis(2-chloroethyl)amino]phenyl]propanoic acid | |
460612 | |
305.2 | |
Melphalan |
180.0°C | |
93% min. (HPLC) | |
C13H18Cl2N2O2 | |
melphalan, alkeran, melfalan, phenylalanine mustard, l-sarcolysine, l-pam, l-sarcolysin, l-phenylalanine mustard, levofalan, p-l-sarcolysin | |
C1=CC(=CC=C1CC(C(=O)O)N)N(CCCl)CCCl | |
305.2 | |
CHEBI:28876 | |
94% |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause cancer.
Suspected of damaging fertility or the unborn child.<br/
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautio
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-726-3
- Melphalan
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H330-Fatal if inhaled.
- H350-May cause cancer.
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H361-Suspected of damaging fertility or the unborn child.
- H300-Fatal if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only