Learn More
Hirschmann™ Measuring flask Class A with pre-packing regulation
Achieve precise standard solutions. Hirschmann™ Measuring flask Class A with pre-packing regulation is standardized to contain or to deliver a definite amount of liquid. Flask has blue graduations.
138.00€ - 226.00€
Product Code | Brand | Graduation Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Graduation Range | Price | Quantity & Availability | |||||
11893212
|
Hirschmann™
2950188 |
323-337 mL |
209.00€
Pack of 2 |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
11823212
|
Hirschmann™
2950165 |
18 to 22 mL |
159.00€
Pack of 2 |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
11692229
|
Hirschmann™
2950184 |
368-382 mL |
210.00€
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11833212
|
Hirschmann™
2950170 |
23 to 27 mL |
159.00€
Pack of 2 |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
11892582
|
Hirschmann™
2950175 |
47 to 53 mL |
171.00€
Pack of 2 |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
11873212
|
Hirschmann™
2950186 |
245 to 255 mL |
195.00€
Pack of 2 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
11803222
|
Hirschmann™
2950189 |
343 to 357 mL |
209.00€
Pack of 2 |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
11813222
|
Hirschmann™
2950190 |
490 to 510 mL |
226.00€
Pack of 2 |
In Stock
Log in to see stock. |
|||||
10421961
|
Hirschmann™
2950191 |
690 to 710 mL |
138.00€
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
Description
- Class A
- Pursuant to the rules of the pre-packing regulation
- Certified conformity
- Blue graduation
- Non-ground, rimmed
- Printed scale on the neck for simple and rapid identification of deviation of actual value from normal value
- Calibrated for IN
Specifications
Yes | |
A |