Learn More
Hirschmann™ Measuring flask Class A with pre-packing regulation
Achieve precise standard solutions. Hirschmann™ Measuring flask Class A with pre-packing regulation is standardized to contain or to deliver a definite amount of liquid. Flask has blue graduations.
Brand: Hirschmann™ 2950184
This item is not returnable.
View return policy
Description
- Class A
- Pursuant to the rules of the pre-packing regulation
- Certified conformity
- Blue graduation
- Non-ground, rimmed
- Printed scale on the neck for simple and rapid identification of deviation of actual value from normal value
- Calibrated for IN
Specifications
Borosilicate Glass (3.3) | |
0.5 mL | |
± 0.15 mL | |
2/Pack | |
Blue enamel color | |
Reusable | |
Non-sterile | |
Round |
368-382 mL | |
260 mm | |
Rim | |
± 0.15 mL | |
Autoclavable | |
Yes | |
A |