missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM7 Mouse, Clone: MCM7/1469, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4176-MSM4-P1
This item is not returnable.
View return policy
Specifications
MCM7 | |
Nuclear | |
MCM7/1469 | |
Unconjugated | |
NP_005907 | |
Mouse | |
88kDa | |
Protein A/G Purified | |
RUO | |
4176 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
MCM7 | |
Recombinant human MCM7 protein fragment (aa195-319) | |
Raji or Ramos cells. Tonsil | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MCM7 Mouse, Clone: MCM7/1469, enQuireBio™ > 100 μg
Spot an opportunity for improvement?Share a Content Correction