Learn More
Masterflex™ L/S™ Platinum-Cured Silicone Pump Tubing, spooled
Reduce scrap with spooled tubing. Masterflex™ L/S™ Platinum-Cured Silicone Pump Tubing features a smooth surface and low protein binding levels, ideal for pharmaceutical, biotechnology, peristaltic pumping and food applications.
Brand: Masterflex™ 96403-36
983.25 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.- Ensure that you always have enough tubing on hand
- Slightly greater clarity
- Preferred by pharmaceutical and biotechnology industries
- Fewer potential leachables
- USP Class VI
- FDA compliant
Specifications
9.7 mm | |
3000 cm | |
-50°C | |
37 | |
3/8 in. | |
610 mm Hg | |
Pump Tubing |
Platinum-cured Silicone | |
Pharmaceutical and biotechnology industries | |
1 Ea. | |
4.8 to 2900 mL/min. | |
40 psi | |
230°C |
Offerte speciali
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92%
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.