Learn More
Watson-Marlow Manifold Tubing - Silicone
Double pump segment manifold tubing — use one, transfer to the other for double life
Brand:  Watson-Marlow 983.0088.000
 View more versions of this product
Product Code. 17877933
  • 342.00€ / Pack of 6
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pump Tubing
0.88 mm
0.03 in.
400DM2 and 400DM3 pump heads and multichannel cartridge
0.09 to 6.4 (four roller pump), 0.08 to 5.3 (eight roller pump)mL/min.
3/100 in.
Silicone
0.8 mm
0.88mm
Orange/Orange