Learn More
Thermo Scientific™ Male Luer-LOK Priming Syringes
Product Code. 10351305 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
Product Type:
GC Autosampler Priming Syringe, Luer-Lok
GC Autosampler Syringe
Gas Tight GC Autosampler Syringe, Luer-Lok
Unit Size:
Each
Product Code. 10351305
Brand: Thermo Scientific™ 365RL581
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
TRUSTED_SUSTAINABILITY
Specifications
Product Type GC Autosampler Priming Syringe, Luer-Lok
Volume (Metric) 50 mL
For Use With (Equipment) Gas Chromatography Systems
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.