Learn More
BINDER Maintenance
One-off maintenance service in accordance with maintenance schedule.
Brand:  BINDER DL20-0615
 View more versions of this product
Product Code. 30274281
  • 891.00€ / Each
Explore more special offers
Description
Description
One-off maintenance service in accordance with maintenance schedule.
  • Visual inspection of mechanical and electrical components, testing of all key functions.
  • Calibration of a test temperature in the center of usable space as specified by the user
  • Without certificate
Specifications
Specifications
BINDER Chambers Series VD
Preventive Maintenance
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER Maintenance > BINDER One Time Maintenance, For Use with Equipment, BINDER Series VDL