Learn More
Cytiva Mains Distribution Lead
Brand: Cytiva 18-1119-06
Specifications
AKTAexplorer | |
Mains Distribution Lead | |
0.5 m |
AKTAexplorer Cytiva | |
1 Pack |
Product Content Correction
Product Title
Cytiva Mains Distribution Lead > Length: 0.5m