Learn More
Hanna Instruments™ Ammonia Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Ammonia Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
156.00€ - 348.00€
Specifications
For Use With (Equipment) | Compatible Hanna benchtop and portable photometers |
---|---|
Quantity | 1 Set |
Type | Water Testing Kit |
Product Code | Brand | For Use With (Application) | No. of Tests | Includes | Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | No. of Tests | Includes | Range | Price | Quantity & Availability | ||
11863683
|
Hanna Instruments™
HI9371901 |
Magnesium and total hardness determination | 100 | Dropper bottles | 0.00 to 2.00mg/L (Mg hardness) and 0.00 to 4.70mg/L (total hardness) |
156.00€
Pack of 100 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
||
11826672
|
Hanna Instruments™
HI9373301 |
Ammonia high range determination | 100 | Dropper bottles | 0.0 to 50ppm ammonium |
246.00€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11748753
|
Hanna Instruments™
HI93764B25 |
Ammonia high range determination | 25 | 25 ready-to-use vials of HI93764B-0 and 1 bottle HI93764-0 Nessler reagent | 0 to 100mg/L ammonia nitrogen |
348.00€
Pack of 25 |
Estimated Shipment: 11-02-2025
Log in to see stock. |
||
Description
These high quality reagents are designed to follow the ASTM Manual of Water and Environmental Technology, D1426, Nessler Method for colorimetric determination of Ammonia. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Pre-made, pre-measured testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each component for traceability
- Tests available for low-range, medium range and high-range determinations
- Kits with 25 ready-to-use single vials of reagent available
Specifications
Compatible Hanna benchtop and portable photometers | |
1 Set | |
Water Testing Kit |
Safety and Handling
- TEST TOTAL HARDNESS
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P390-Absorb spillage to prevent material damage.
- P391-Collect spillage.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide, EDTA tetrasodium salt