Learn More
Hanna Instruments™ Ammonia Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Ammonia Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
Brand: Hanna Instruments™ HI93764B25
This item is not returnable.
View return policy
Description
These high quality reagents are designed to follow the ASTM Manual of Water and Environmental Technology, D1426, Nessler Method for colorimetric determination of Ammonia. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Pre-made, pre-measured testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each component for traceability
- Tests available for low-range, medium range and high-range determinations
- Kits with 25 ready-to-use single vials of reagent available
Specifications
Ammonia high range determination | |
25 ready-to-use vials of HI93764B-0 and 1 bottle HI93764-0 Nessler reagent | |
1 | |
0 to 100mg/L ammonia nitrogen | |
Water Testing Kit |
Compatible Hanna benchtop and portable photometers | |
25 | |
1 Set | |
D1426-92 Nessler |