missing translation for 'onlineSavingsMsg'
Learn More

Magnesium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11319438
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Quantity 50 g
Percent Purity 99.95%
Odor Odorless
Weight ≈0.35g/slug
Form Slug
Assay (metals basis)
Chemical Name or Material Magnesium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI09Q34 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043298.30,043299.30,043298.09,043298.18,043299.18
Lot NumberI04S041 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043298.30,043299.30,043298.09,043298.18,043299.18
Lot NumberZ21G049 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043298.30,043299.30,043298.09,043298.18,043299.18
Lot NumberF03N15 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043298.30,043299.30,043298.09,043298.18,043299.18
Lot NumberB18M12 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043298.30,043299.30,043298.09,043298.18,043299.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Magnesium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.