Learn More
Magnesium chloride, anhydrous, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Melting Point | 714°C |
Color | White to Yellow |
Physical Form | Crystals |
Quantity | 1 kg |
Assay Percent Range | 99% |
Solubility Information | Highly soluble in water with evolution of heat. Slightly soluble in acetone and pyridine. |
IUPAC Name | magnesium(2+) dichloride |
Formula Weight | 95.22 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P12L028 | Certificate of Analysis | 14/03/2025 | 012315.A1,012315.C4 |
Y22K040 | Certificate of Analysis | 21/01/2025 | 012315.A1,012315.C4 |
Y22K038 | Certificate of Analysis | 16/12/2024 | 012315.A1,012315.C4 |
Y22K041 | Certificate of Analysis | 16/12/2024 | 012315.A1,012315.C4 |
S19K025 | Certificate of Analysis | 27/06/2024 | 012315.A1,012315.C4 |
Promotions
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.