missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MabCaptureC Affinity Matrix

Thermo Scientific™ MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast.

4755.00€ - 92300.00€

Specifications

Binding Capacity >50 g IgG/L
Ligand Engineered rProtein A
Matrix Agarose-based, epoxide-activated
Particle Size 75 ±10 μm
Target CH2-CH3 interface Fc-region
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30019889
View Documents
Thermo Scientific™
1963662250
250 mL
4755.00€
250mL
Please to purchase this item. Need a web account? Register with us today!
30019838
View Documents
Thermo Scientific™
196366201L
1 L
9490.00€
1 L
Please to purchase this item. Need a web account? Register with us today!
30019782
View Documents
Thermo Scientific™
196366205L
5 L
47375.00€
5 L
Please to purchase this item. Need a web account? Register with us today!
30274933
View Documents
Thermo Scientific™
196366210L
10 L
92300.00€
10 L
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

>50 g IgG/L
Agarose-based, epoxide-activated
CH2-CH3 interface Fc-region
Agarose Bead
Engineered rProtein A
75 ±10 μm
Monoclonal antibody purification
Videos
SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MabCaptureC Affinity Matrix

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.