Learn More
Emma Safety Footwear Lukas XXD6T Safety Shoes
An orthopedic version of the standard Lukas is designed, dedicated and created for those with slightly broader than normal feet or orthopedic requirements.
114.00€
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
|---|---|
| Closure Type | Laces |
| Color | Black |
| Design | Lukas XXD6T |
| Liner Material | Hydro-Tec™ Sanitized Silver |
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather
- Steel toe cap
- PU outer toe cap
- Extremely resilient reinforced sole
- Hydro-Tec™ Sanitized Silver
Specifications
| EN ISO 20345:2011 | |
| Black | |
| Hydro-Tec™ Sanitized Silver | |
| Steel |
| Laces | |
| Lukas XXD6T | |
| Leather |
Product Title
Emma Safety Footwear Lukas XXD6T Safety Shoes