Learn More
Fisherbrand™ LN2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Fisherbrand™ Isotemp LN2 Backup Systems for Ultra-Low Temperature Freezers.
3410.00€
Specifications
Description Factory Installed
Temperature Range -80°C
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
17858010
View Documents Promotion Details
Fisherbrand™
332353G03
Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer
3410.00€
Each
Estimated Shipment: 30-06-2025
to see stock.
Add to basket
17898010
View Documents Promotion Details
Fisherbrand™
332353G07
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer
3410.00€
Each
Estimated Shipment: 30-06-2025
to see stock.
Add to basket
Description
Description
  • Maintains temperature down to -80°C with liquid nitrogen
Specifications
Specifications
Factory Installed
-80°C
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ LN2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers