missing translation for 'onlineSavingsMsg'
Learn More
Lithium 2,4-pentanedionate, 99.5%, Thermo Scientific Chemicals Product Code.: 11337298

Lithium 2,4-pentanedionate, 99.5%, Thermo Scientific Chemicals

Product Code. 11337298
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11337298

Brand: Thermo Scientific Alfa Aesar 030529.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18115-70-3
Molecular Formula C5H7LiO2
Molecular Weight (g/mol) 106.05
MDL Number MFCD00013496
InChI Key JTEOOCAGEXVCBQ-LNKPDPKZSA-M
Synonym Lithium acetylacetonate
IUPAC Name lithium(1+) (2Z)-4-oxopent-2-en-2-olate
SMILES [Li+].C\C([O-])=C\C(C)=O

Specifications

Melting Point ∼250°C (decomposition)
Color White
Quantity 50 g
Beilstein 4575087
Sensitivity Hygroscopic
Formula Weight 106.05
Percent Purity 99.5%
Physical Form Crystalline
Chemical Name or Material Lithium 2,4-pentanedionate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN11F028 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog Number030529.18,030529.30,030529.09
Lot NumberU18J046 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog Number030529.18,030529.30,030529.09
Lot NumberW24E067 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number030529.18,030529.30,030529.09
Lot NumberI12R032 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number030529.18,030529.30,030529.09
Lot NumberM26E038 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number030529.18,030529.30,030529.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium 2,4-pentanedionate, 99.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.