missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semadeni™ Lid for Color-Coded Plastic Beakers
Click to view available options
For Use With (Equipment):
Color-Coded Plastic Beakers
Description
- Natural in color
- Snap-on closure
Specifications
Specifications
Material | Polypropylene |
For Use With (Equipment) | Color-Coded Plastic Beakers |
Quantity | 100 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Semadeni™ Lid for Color-Coded Plastic Beakers >
Spot an opportunity for improvement?Share a Content Correction