Learn More
Brady™ Laminated Polyester Warning Sign
Brand:  Brady™ 250201
 View more versions of this product
Product Code. 10000674
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Specifications
ISO 7010
1 Ea.
Laminated Polyester
100 mm
100 mm
Triangular
Warning Sign
Polyester
Permanent Acrylic
Pipemarkers and safety signs
0.024 mm
-40°C to 110°C
Warning of Flammable Materials
Black on Yellow