Learn More
Brady™ Laminated Polyester Label Markers
Laminated Polyester Label Markers feature a surface printed white polyester film with a permanent, acrylic based, pressure sensitive adhesive and over-laminated with a clear polyester film.
Brand:  Brady™ 271389
 View more versions of this product
Product Code. 11745563
  • 340.00€ / 1 roll
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used for pipe markers and safety signs.
  • B-7541 gives excellent adhesion to low surface energy surfaces, such as polypropylene and ABS, as well as on most powder coatings.
  • Standard ink colors are black, red (RAL 3001-GL), yellow (RAL 1003-GL), blue (RAL5005-GL) and green (RAL 6032-GL).
Specifications
Specifications
PMF-EAUINCENDIE-RD4-RLL090
Permanent Acrylic
Gloss
Polyester
Eau incendie
French
220 Pk.
-40°C to +110°C
0.099 mm
33 m x 100 mm x 150 mm
33 m
100 mm
150 mm
3 in.
Pipe Markers - Eau incendie
White on Red
Roll
Pipe Marker
Pipe and valve marking
220 Markers/Roll
-40°F to +230°F
0.0039 in.
33 m x 100 mm
108.27 ft.
3.94 in.
5.91 in.
13 mm
70 mm