Learn More
3M™ Laminate Over Boot
Brand: 3M™ 7000088638
345.47 EUR valid until 2025-06-08
Use promo code "24131" to get your promotional price.
Description
Specifications
CE Category 1 | |
Laminate | |
Overboot |
White | |
Universal |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now