Learn More
Labconco™ Vacuum Tubing Kit for CentriVap™ System and RapidVap™
Vacuum Tubing Kit for use with Labconco CentriVap Vacuum Concentrators and RapidVap Evaporators.
Brand:  Labconco™ 7814500
Product Code. 10409543
  • 241.00€ / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Vacuum Tubing Kit for CentriVap Concentrators and RapidVap Evaporators includes two each 1/2 in. ID vacuum tubing, 48 in. length, and 4 tubing clamps.
Specifications
Specifications
CentriVap™ system
Vacuum Tubing Kit
6 lb.
2.7 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labconco™ Vacuum Tubing Kit for CentriVap™ System and RapidVap™ > Vacuum Tubing Kit For CentriVap System