Learn More
Labconco™ SpillStopper™ Work Surface for 6ft. Protector™XStream
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
2840.00€
Specifications
For Use With (Equipment) | Protector XStream Fume Hood (72 in.) |
---|---|
Product Type | Work Surface |
Material | Epoxy Resin |
For Use With (Application) | Laboratory Fume Hood Work |
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
13141144
|
Labconco™
9503610 |
Left Rear | N/A | N/A | N/A | |||||
Description
- Dished and contoured to conform to interior liner of Protector Laboratory Hoods
- Front edge with large radius to aerodynamically direct airflow in hood
- Pre-drilled holes for venting and plumbing passthroughs
- Includes left rear cupsink cutout
Specifications
Protector XStream Fume Hood (72 in.) | |
Work Surface | |
Epoxy Resin | |
Laboratory Fume Hood Work |
Product Title
Labconco™ SpillStopper™ Work Surface for 6ft. Protector™XStream