missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ MART-1/Melan-A (Melanoma Marker) Ab-4, Mouse Monoclonal Antibody
Epredia™ MART-1/Melan-A (Melanoma Marker) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia™ MS-799-P
580.50 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. MART-1 is present in melanosomes and endoplasmic reticulum.Host Species: Mouse
Clone: A103
Isotype: IgG1
Species Reactivity: Human and Dog. Others not tested.
Epitope: Not determined
Immunogen: Recombinant MART-1 protein
Molecular Weight: 18kDa
Positive Control: CAC1 melanoma cells and Melanomas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
Specifications
MART-1/Melan-A (Melanoma Marker) Ab-4 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Cancer and Tumor Biology | |
Canine, Human | |
IgG1 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
A103 | |
Unconjugated | |
Recombinant MART-1 protein | |
IVD | |
Primary | |
Concentrated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction