Learn More
OHAUS™ Lab Lift
OHAUS™ Lab Lift provides secure support for holding glassware at elevated positions.
Brand: OHAUS™ 30400004
339.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
Specifications
Lab Lift | |
Clamp, Support, Lab-Lift, CLR-LBLFTS025 | |
254 x 254 (L x W) Platform |
Stainless Steel | |
89 to 330 mm |
Product Content Correction
Product Title
OHAUS™ Lab Lift > Material: Stainless Steel; Platform Size: 254 x 254 (L x W)