missing translation for 'onlineSavingsMsg'
Learn More
L-Valine, Cell Culture Reagent, Thermo Scientific Chemicals Product Code.: 15405249

L-Valine, Cell Culture Reagent, Thermo Scientific Chemicals

Product Code. 15405249
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
This item is not returnable. View return policy

Product Code. 15405249

Brand: Thermo Scientific Alfa Aesar J62943.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-18-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064220
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
Synonym l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latinShow MoreShow Less
PubChem CID 6287
ChEBI CHEBI:16414
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@H](N)C(O)=O

Specifications

Color White
Beilstein 1721136
Merck Index 14,9909
Quantity 25 g
Odor Odorless
Formula Weight 117.15
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Valine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS27I052 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog NumberJ62943.22,J62943.A1,J62943.03,J62943.14,J62943.06
Lot NumberQ19H073 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ62943.22,J62943.A1,J62943.03,J62943.14,J62943.06
Lot NumberX22H041 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ62943.22,J62943.A1,J62943.03,J62943.14,J62943.06
Lot NumberU21G009 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ62943.22,J62943.A1,J62943.03,J62943.14,J62943.06
Lot NumberT06H006 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ62943.22,J62943.A1,J62943.03,J62943.14,J62943.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Valine, Cell Culture Reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.