Learn More
Medicom™ KOLMI Waterproof Overshoes with Non-slip SafeFeet SkidGuard Sole
Click to view available options
KOLMI Waterproof Overshoes with Non-slip SafeFeet SkidGuard Sole; Size: X-Large
Description
- Made of non-woven spunbonded 100% polypropylene
- Glue sealing
- Elastic, latex-free ankle
- Supple and slip resistant
- Anti-slip 60 μm white sole
Specifications
Specifications
Size | X-Large |
Material | Polypropylene |
Latex Free | Yes |
Coating | None |
Color | White, Blue |
Disposable | Single-use |
Sterility | Non-sterile |
Closure Type | Elastic Ankle |
Sole Type | Anti-slip |
Resistance | Water |
Show More |
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Medicom™ KOLMI Waterproof Overshoes with Non-slip SafeFeet SkidGuard Sole >