Learn More
LNI Swissgas Kit Service NG SIRIO 5000 HP
Kit service 16000 h NG SIRIO 5000 HP.
Brand:  LNI Swissgas 6930.00.191
Product Code. 17862219
  • 2180.00€ / Each
Explore more special offers
Description
Description
  • To be carried out by trained service engineer every 16000 hours or 4 years.
Specifications
Specifications
PM Kit
Product Title
LNI Swissgas Kit Service NG SIRIO 5000 HP > LNI Swissgas Kit Service