Learn More
HMC Europe Vertical Autoclave Baskets
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
Brand: HMC Europe 50009
This item is not returnable.
View return policy
Description
For use with the HMC-HG and HMC-HV autoclaves.Specifications
15.35 in. | |
29 gal. | |
HV 110 autoclave | |
380 mm | |
Stainless Steel |
390 mm | |
110 L | |
14.96 in. | |
Stackable basket | |
390 x 380 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Product Title
HMC Europe Vertical Autoclave Baskets > HMC-HV 110 L; Basket; Dia. x H: 390 x 380mm