Learn More
Kimberly-Clark Professional™ Kimtech™ A5 Sterile Cleanroom Coveralls
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
Assured Compliance:
- PPE Cat III according to Regulation (EU) 2016/425 (EU)
- Type 5 and 6 certified
- Certificate of conformance and sterility by lot available online
- Suitable for use in EU GMP ISO 5 Class A sterile cleanroom environments and laboratories
- Featuring advanced Clean-Don™ technology for fast, simple and compliant aseptic donning
- Made from low-lint breathable SMS fabric with a cloth-like feel
- Contamination protection is ensured through high quality sterility assured material with secure ties
- Individually vacuum-packed and efficiently double-bagged to save space
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Specifications
Product Type | Cleanroom Coveralls |
Size | 4X-Large |
Material | SMS |
Color | White |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Collared |
Hood Type | Unhooded |
Show More |
Videos
-
KIMTECH - Reusable Apparel
Documents
Scientific Resources
Brochures
KIMTECH - Are you dressed for success?
File size: 1.6 MB
Product Information
Product Information
KCP015 4518.02 F 03.09 KIMTECH PURE A5 Cleanroom Apparel data sheet
File size: 868 kB
Promotions
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Days
5Hours
11Minutes
56Seconds
18
Product Content Correction
Product Title
Kimberly-Clark Professional™ Kimtech™ A5 Sterile Cleanroom Coveralls >