missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Center Well for Incubation Flask

The top stopper for the Kontes™ sidearm incubation flask has an off-center hole for the insertion of the disposable polypropylene center well. The center well may be cut off when transferred to a liquid scintillation counting vial.

Brand:  DWK Life Sciences 882320-0000

Product Code. 11805824

  • 371.00€ / Pack of 144
Estimated Shipment: 30-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Kontes™ sidearm incubation flask is used for metabolic studies of biological materials. It is particularly useful in measuring C14O2 yields following incubation, and is also suitable for in vitro incubations, insulin bio-assays, persulfate oxidations, and the “distillation” of any volatile acid or base.

  • Since either an acid or alkali is introduced at the end of the reaction, it is possible to use this easy system with either a phosphate or bicarbonate buffer
  • Sidearm flask enables syringe needle entry without disturbing established conditions
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Top stoppers, sidearm stoppers and center wells are available separately
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.39 in.
For metabolic studies of biological materials
144
1 cm
Polypropylene
For incubation flask
Center well
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Kimble™ Kontes™ Center Well for Incubation Flask > Center Well, Diameter: 1mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.