Learn More
DWK Life Sciences Kimble™ Kontes™ Filtration Assemblies: Flasks With Side Arm, No. 2 Hose connection
Borosilicate glass flask with side arm, No. 2 hose connection.
Brand:  DWK Life Sciences 953760-4000
 View more versions of this product
Product Code. 11883173
  • 443.00€ / Each
Estimated Shipment: 27-10-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DWK Life Sciences Kimble Kontes Filtration Assemblies: Flasks With Side Arm, No. 2 Hose connection, Connector Types: No. 2 Hose Connection, Borosilicate Glass, Description: Filtration Flask
Order Info
Does not include rubber stopper and hose
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No. 2 Hose Connection
1/Cs.
Filtration Flask
Borosilicate Glass
8
4000 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ Filtration Assemblies: Flasks With Side Arm, No. 2 Hose connection > Flask filter; Capacity: 4000mL; No 8 Stpr Joint