Description
Chemsplash Jet Spray Overboot with Taped Seams Type PB (3-B)
- 88 GSM Chemsplash Jet Spray Fabric
- Anti-Slip Sole and Anti-Static Strips
- Low Linting
- Elasticated Top and Ankles
- Taped Seams
- PFAS Free
Description: The Chemsplash Jet Spray Overboot with Taped Seams Type PB (3-B) is made from 88 GSM Chemsplash Jet Spray fabric. It is highly resistant to chemical splash and dust, providing a strong protective barrier. The overboot features a PVC anti-slip and anti-static sole, making it ideal for safe and secure movement in environments with potential hazards. The elasticated top and ankles ensure a secure fit, while the taped seams provide added protection. This overboot is low-linting, helping maintain a clean environment. The Chemsplash Accessories range is designed to protect both the wearer and the environment, offering maximum comfort and wearability.
- Environmental benefits include:
- Recycled Content
Specifications
Specifications
Color | Yellow |
Material | Polyethylene Laminate |
Size | Medium/Large |
Waterproof | Yes |
Product Type | Controlled Environments Boots |