missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ IWP 4
Potent inhibitor of Wnt/β-catenin signaling
278.00€ - 1150.00€
Specifications
Purity | >98% |
---|---|
Product Type | IWP 4 |
Molecular Weight (g/mol) | 496.62 |
Inhibitors | IWP 4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732428
|
Tocris Bioscience™
5214/10 |
10 mg |
278.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18760159
|
Tocris Bioscience™
5214/50 |
50 mg |
1150.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent inhibitor of Wnt/β-catenin signaling (IC50 = 25 nM). Has minimal effect on Notch and Hedgehog signaling pathways. Induces differentiation of cardiomyocytes from human ESCs and iPSCs.Specifications
>98% | |
IWP 4 | |
496.62 | |
IWP 4 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ IWP 4