missing translation for 'onlineSavingsMsg'
Learn More

IVD Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-18798MFV610

Product Code. 18586189

  • 481.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

IVD Polyclonal antibody specifically detects IVD in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IVD
Polyclonal
ELISA
ACAD2, EC 1.3.99, EC 1.3.99.10, FLJ12715, FLJ34849, isovaleryl-CoA dehydrogenase, mitochondrial
Produced in rabbits immunized with purified, recombinant Human IVD (Accession#: AAH17202.1; His30-His423)
0.1 mL
Stem Cell Markers
3712
Store at 4C in the dark.
IgG
ELISA
mFluor Violet 610 SE
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IVD Antibody, mFluor Violet 610 SE, Novus Biologicals™ > 0.1 mL; mFluor Violet 610 SE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.