missing translation for 'onlineSavingsMsg'
Learn More

Ionisation Buffer Solution for AAS and ICP, Cesium 2% Cs in HNO3 1%, Honeywell Fluka™

248.33€ - 580.00€

Specifications

Chemical Name or Material Ionisation buffer Solution for AAS and ICP
Density 1.01 g/cm3
pH <2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16644454
View Documents
Honeywell Fluka
E4300-100ML
100 mL
248.33€
100mL
Please to purchase this item. Need a web account? Register with us today!
16654454
View Documents
Honeywell Fluka
E4300-500ML
500 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
Description

Description

Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

Specifications

Specifications

Ionisation buffer Solution for AAS and ICP
1.01 g/cm3
<2
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 760

SDS
Documents

Documents

Product Certifications