Learn More
Ionisation Buffer Solution for AAS and ICP, Cesium 2% Cs in HNO3 1%, Honeywell Fluka™
Brand:  Honeywell Fluka E4300-500ML
 View more versions of this product
Product Code. 16654454
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ionisation buffer Solution for AAS and ICP
500 mL
1.01 g/cm3
<2