missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Iodonitrotetrazolium violet, 95% Product Code.: 11415986

Thermo Scientific Chemicals Iodonitrotetrazolium violet, 95%

Product Code. 11415986
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11415986

Brand: Thermo Scientific Chemicals B22301.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for dehydrogenase determination

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146-68-9
Molecular Formula C19H13ClIN5O2
Molecular Weight (g/mol) 505.70
MDL Number MFCD00011961,MFCD00149999
InChI Key JORABGDXCIBAFL-UHFFFAOYSA-M
Synonym iodonitrotetrazolium chloride, iodonitrotetrazolium, iodonitrotetrazolium purple, iodonitrotetrazolium violet, 2-4-iodophenyl-3-4-nitrophenyl-5-phenyltetrazolium chloride, iodonitro tetrazolium, int, p-iodonitrotetrazolium violet, int chemical, 2-p-iodophenyl-3-p-nitrophenyl-5-phenyltetrazolium chlorideShow MoreShow Less
PubChem CID 64957
ChEBI CHEBI:75421
IUPAC Name 2-(4-iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;chloride
SMILES [Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1

Specifications

Melting Point ∼231°C (decomposition)
Quantity 5 g
Beilstein 4093224
Sensitivity Light sensitive
Formula Weight 505.70 (Anhydrous)
Percent Purity 95%
Chemical Name or Material Iodonitrotetrazolium violet

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247513 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberB22301.03,B22301.14,B22301.06
Lot Number10229113 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog NumberB22301.03,B22301.14,B22301.06
Lot Number10243224 Certificate TypeCertificate of Analysis Date09/09/2022 Catalog NumberB22301.03,B22301.14,B22301.06
Lot Number10157849 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22301.03,B22301.14,B22301.06
Lot Number10211755 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22301.03,B22301.14,B22301.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Iodonitrotetrazolium violet, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.