missing translation for 'onlineSavingsMsg'
Learn More
Iodic acid, 99% Product Code.: 11418990
  • ONLINE EXCLUSIVE PRICE
    587.00€
    584.02€ / 1000g
    Save 2.98€
    0.51% Off

Iodic acid, 99%

Product Code. 11418990
1000 g, 1000g
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
50g
This item is not returnable. View return policy

Product Code. 11418990

Brand: Thermo Scientific Alfa Aesar A11925.0B

  • ONLINE EXCLUSIVE PRICE
    587.00€
    584.02€ / 1000g
    Save 2.98€
    0.51% Off
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-68-5 | ILiO3 | 181.84 g/mol

Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.

Solubility
Soluble in water and methanol.

Notes
Light sensitive. Incompatible with reducing agents, alcohols and organic materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-68-5
Molecular Formula ILiO3
Molecular Weight (g/mol) 181.84
MDL Number MFCD00011348
InChI Key FZAXZVHFYFGNBX-UHFFFAOYSA-M
PubChem CID 24345
ChEBI CHEBI:24857
IUPAC Name lithium(1+) iodate
SMILES [Li+].[O-][I](=O)=O

Specifications

Melting Point 110°C (decomposition)
Density 4.629
Odor Odorless
Sensitivity Light sensitive
Quantity 1000 g
UN Number UN3085
Merck Index 14,5012
Synonym unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
Solubility Information Soluble in water and methanol.
Formula Weight 175.91
Percent Purity 99%
Chemical Name or Material Iodic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10251774 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeA11925.18,A11925.30,A11925.0B
Lot Number10249415 Certificate TypeCertificate of Analysis Date19/01/2024 Product CodeA11925.18,A11925.30,A11925.0B
Lot Number10242870 Certificate TypeCertificate of Analysis Date17/08/2022 Product CodeA11925.18,A11925.30,A11925.0B
Lot NumberF03X028 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA11925.18,A11925.30,A11925.0B
Lot Number10199919 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA11925.18,A11925.30,A11925.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodic acid, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.