missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ DNase/RNase-Free Distilled Water
GREENER_CHOICE

Designed for use in all molecular biology applications

15.00€

Specifications

Grade Molecular Biology
Packaging Bottle
Treatment(s) Not DEPC-Treated
Purification Method Membrane-Filtered
Product Line UltraPure
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15657708
GREENER_CHOICE
View Documents
Invitrogen™
10977015
500 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
Description

Description

  • UltraPure DNase/RNase-Free Distilled Water is 0.1-μm membrane-filtered and tested for DNase and RNase activity
  • Performance and quality testing: No DNase, RNase, or protease activity detected. Our distilled water system is routinely monitored for compliance with current USP monograph test requirements for Water for Injection (WFI)
  • Sustainable packaging

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Molecular Biology
Not DEPC-Treated
UltraPure
Bottle
Membrane-Filtered
Videos
Safety and Handling

Safety and Handling

Recommended Storage : • 1 bottle of UltraPure DNase/RNase-Free Distilled Water (500 mL/bottle)

Store at room temperature.

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ UltraPure™ DNase/RNase-Free Distilled Water

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.