Learn More
Invitrogen™ T-REx™-HeLa Cell Line
Stably express the tetracycline repressor protein
Brand: Invitrogen™ R71407
Specifications
1 x 107 cells are supplied frozen in 1 ml 90% complete medium and 10% DMSO. Store in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored. | |
Human | |
T-REx™ |
T-REx-HeLa Cell Line | |
1 x 107 | |
1 x 107 cells |
User Guide: Growth and Maintenance of T-REx Cell Lines
Version: 5.0
File size: 275 kB
Product Content Correction
Product Title
Invitrogen™ T-REx™-HeLa Cell Line > 3 x 106 cells; HeLa cell line