Learn More
Invitrogen™ pBAD TOPO™ TA Expression Kit
Specifically designed for one-step cloning and regulated prokaryotic expression of Taq-amplified PCR products
Brand:  Invitrogen™ K430001
 View more versions of this product
Product Code. 10115092
  • 1206.00€ / Each
Explore more special offers
Includes: Each pBAD TOPO TA Expression Kit contains two boxes and the LMG194 E. coli stab. The pBAD TOPO TA box contains all of the reagents necessary for PCR (except Taq polymerase), including 200ng of topoisomerase I-activated pBAD-TOPO vector, sterile water, dNTPs, 10X PCR buffer, salt solution, control template and primers, 20% L-arabinose, pBAD primers for sequencing or PCR screening, and an expression control plasmid. The other box contains all of the reagents necessary for transformation, including single-use 50μL aliquots of TOP10 Chemically Competent E. coli, S.O.C. medium, and a supercoiled control plasmid.
Description
Description
  • Linearized, topoisomerase I-activated vector for 5-minute cloning of Taqamplified PCR products
  • araBAD promoter for tightly regulated expression in E. coli V5 epitope tag for detection with Anti-V5 Antibody
  • C-terminal polyhistidine (6xHis) tag for purification using nickel-chelating resin and detection with Anti-His(C-term) Antibody
  • Enterokinase cleavage site for removal of N-terminal leader peptide
Bacterial Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Inducible
araBAD
None
Ampicillin (AmpR)
EK (Enterokinase) Recognition Site
Cell-Based Expression
TOPO-TA
pBAD
Arabinose
TA Expression Kit
Each pBAD TOPO™ TA Expression Kit contains two boxes and the LMG194 E. coli stab. The pBAD TOPO™ TA box contains all of the reagents necessary for PCR (except Taq polymerase), including 200 ng of topoisomerase I-activated pBAD-TOPO™ vector, sterile water, dNTPs, 10X PCR buffer, salt solution, control template and primers, 20% L-arabinose, pBAD primers for sequencing or PCR screening, and an expression control plasmid. Store at -20°C. The One Shot™ box contains all of the reagents necessary for transformation, including single-use 50 μl aliquots of One Shot™ TOP10 Chemically Competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Store One Shot™ cells at -80°C. Store the LMG194 E. coli stab at 2 - 8°C. All reagents are guaranteed stable for 6 months when properly stored.
LMG194, TOP10
E. coli
His Tag (6x), V5 Epitope Tag
20 Reactions
TOPO™