Learn More
Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
Brand:  Hanna Instruments™ HI95771-01
 View more versions of this product
Product Code. 11756003
  • 85.10€ / Pack of 100
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determinations of total and/or free chlorine. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
  • Reagent sets available for: Free and total chlorine (high range), Free chlorine (ultra low range), Total chlorine (ultra high range, ultra low range, undetermined range)
Specifications
Specifications
Chlorine Ultra High Range determination
Individual packets
1 Set
Standard Methods, 20th edition, 4500-Cl
Compatible Hanna benchtop and portable photometers
100
0 to 500mg/L (ppm) total chlorine
Water Testing Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing > Chlorine Ultra High Range determination; 100 Tests