Learn More
Elma™ Insert Covers
Insert cover contains two rubber rings and is designed for hanging cleaning cups or glasses in the cleaning basket.
Brand:  Elma™ 1111921
 View more versions of this product
Product Code. 17831347
  • 82.00€ / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Elma ultrasonic units can be operated with or without a cover
  • For more efficient operation, Elma offers various insert covers made of robust and easy-to-clean stainless steel
  • Covers make it possible to clean objects made of different materials simultaneously – but separately
  • If used correctly, all Elma inserts will help to prevent damage to the ultrasonic tank and thus extend the service life of the ultrasonic cleaner
  • Allows cleaning with aggressive fluids
  • Prolongs the service life of the unit
Specifications
Specifications
Lid
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic S, Elmasonic Med, Elmasonic Select, Elmasonic EASY
180
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Elma™ Insert Covers > Insert Covers; Model: 180